Acetonitril-D3 99.9 atom % D for NMR spectroscopy

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Cyanomethane-D3, Acetonitrile-d3, Ethannitrile-D3, Trideuteroacetonitrile, Methyl-d3 cyanide, Methyl cyanide-D3, Acetonitrile-[²H₃]

351390100
ACRO351390100EA 735 DKK
ACRO351390100
Acetonitril-D3 99.9 atom % D for NMR spectroscopy
Acetonitril-D3
Formel: D₃CCN
Molekylvægt: 44.03 g/mol
Kogepunkt: 80.7
Smeltepunkt: –46 °C
Massefylde: 0.844 g/cm³ (20 °C)
Flammepunkt: 2 °C (closed cup)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00001881
CAS-nummer: 2206-26-0
EINECS: 218-616-5
UN nummer: 1648
ADR: 3 II

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless Liquid
Water ≤0.03 % (Coulometric)
D-Enrichment ≥99.85 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR