(R)-(+)-Hydroxypyrrolidin 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: (R)-Pyrrolidin-3-ol, (R)-(+)-3-Pyrrolidinol, (R)-(+)-3-Hydroxypyrrolidine

344570010 344570050
ACRO344570010EA 467 DKK
ACRO344570010 ACRO344570050
(R)-(+)-Hydroxypyrrolidin 98%
(R)-(+)-Hydroxypyrrolidin
Formel: C₄H₉NO
Kogepunkt: 108…110 °C (8 mmHg)
Flammepunkt: >110 °C
Opbevaringstemperatur: Fryser
MDL Number: MFCD00145220
CAS-nummer: 2799-21-5

Bestil nu

Specifikationer Testresultater

Appearance Clear yellow to orange-brown Viscous liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.4870 to 1.4910 (20°C, 589 nm)
GC ≥97.5 %
Specific optical rotation +4.5° to +7.5° (20°C, 589 nm) (c=3.5, CH3OH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR