Dess-Martin periodinane 15% (w/w) in dichloromethane

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1,1-Dihydro-1,1,1-triacetoxy-1,2-benziodoxol-3(1H)-one, DMP, (1,1,1-Triacetoxy)-1,1-dihydro-1,2-benziodoxol-3(1H)-one, Acetic acid 1,1-diacetoxy-3-oxo-1l5-ioda-2-oxa-indan-1-yl ester

333110500 333112500 333110100
ACRO333110500EA 1920 DKK
ACRO333110500 ACRO333112500 ACRO333110100
Dess-Martin periodinane 15% (w/w) in dichloromethane
Dess-Martin periodinane

Appearance: Colorless to light yellow Solution

Formel: C₁₃H₁₃IO₈
Massefylde: 1,362 g/cm³
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00130127
CAS-nummer: 87413-09-0
UN nummer: 1593
ADR: 6.1,III

Bestil nu

Specifikationer Testresultater

Appearance Colorless to light yellow Solution (May develop some turbidity or precipitate)
Titration Iodometric 10 to 15 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR