Acrylamid-Bisacrylamid (19:1) 40% mix in water for biochemistry

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Acrylamid-Bis

330201000 330205000
ACRO330201000EA 434 DKK
ACRO330201000 ACRO330205000
Acrylamid-Bisacrylamid (19:1) 40% mix in water for biochemistry
Acrylamid-Bis (32:1)
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00080848
CAS-nummer: 79-06-1
ADR: 6.1,III

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless Solution
UV at 290 nm A: ≤0.1
Acrylic acid (HPLC) ≤0.001 %
Conductivity ≤3 µS/cm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR