3,3',5,5'-Tetramethylbenzidin Easy, liquid substrate

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine

328030300 328031000
ACRO328030300EA 400 DKK
ACRO328030300 ACRO328031000
3,3',5,5'-Tetramethylbenzidin Easy, liquid substrate
3,3',5,5'-Tetramethylbenzidin
Formel: C₁₆H₂₀N₂
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00007748
CAS-nummer: 54827-17-7

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless to yellow-green to light blue Solution
Absorbance (Background level)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR