Tin 99.5%, powder -100 mesh

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

317811000 317815000
ACRO317811000EA 400 DKK
ACRO317811000 ACRO317815000
Tin 99.5%, powder -100 mesh
Tin
Formel: Sn
Kogepunkt: 2270 °C (760 mmHg)
Massefylde: 7,3 g/cm³
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00133862
CAS-nummer: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587

Bestil nu

Specifikationer Testresultater

Appearance Grey Powder
Total trace metal impurities ≤0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR