Methyl (4-fluorophenyl)acetate 99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Methyl 2-(4-fluorophenyl)acetate

304670250
ACRO304670250EA 973 DKK
ACRO304670250
Methyl (4-fluorophenyl)acetate 99%
Methyl (4-fluorophenyl)acetate
Formel: C₉H₉FO₂
Molekylvægt: 168.17 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00800608
CAS-nummer: 34837-84-8

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4860 to 1.4880 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR