Chloromethyl butyrate 99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Chloromethylbutyrate, Butyric Acid Chloromethyl Ester

295580250 295580050
ACRO295580250EA 1030 DKK
ACRO295580250 ACRO295580050
Chloromethyl butyrate 99%
Chloromethyl butyrate

Appearance: Clear colorless to light yellow Liquid

Formel: C₅H₉ClO₂
Kogepunkt: 150 °C (993mbar)
Massefylde: 1,074 g/cm³
Flammepunkt: 55 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00216961
CAS-nummer: 33657-49-7
UN nummer: 2924
ADR: 3,III

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4190 to 1.4220 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR