Terephthalaldehyde mono(diethyl acetal) 97% stabilised

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4-(Diethoxymethyl)benzaldehyde

294631000 294630250
ACRO294631000EA 2360 DKK
ACRO294631000 ACRO294630250
Terephthalaldehyde mono(diethyl acetal) 97% stabilised
Terephthalaldehyde mono(diethyl acetal)

Appearance: Clear colorless to yellow Liquid

Formel: C₁₂H₁₆O₃
Kogepunkt: 89…93 °C (7 mmHg)
Massefylde: 1,04 g/cm³
Flammepunkt: >112 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00010217
CAS-nummer: 81172-89-6

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5055 to 1.5085 (20°C, 589 nm)
GC ≥96.0 %
Stabilizer 0.1 % triethylamine


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR