Pyridin 99+% for analysis

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Azabenzene, Azine

290860010 290860025 290860100 290861000
ACRO290860010EA 1290 DKK
ACRO290860010 ACRO290860025 ACRO290860100 ACRO290861000
Pyridin 99+% for analysis
Pyridine
Formel: C₅H₅N
Kogepunkt: 115…116 °C (760 mmHg)
Smeltepunkt: –42 °C
Massefylde: 0,978 g/cm³
Flammepunkt: 17 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011732
CAS-nummer: 110-86-1
UN nummer: 1282
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f9c5934940218027dae0d80a1f98b6d5179c0a8&vid=9f2ef46a26e488d366e82869b3acb96652b5dab1&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Clear liquid
Water ≤0.1 %
GC ≥99.0 %
Color scale ≤10 APHA
Non-volatile matter ≤0.002 %
Ammonia (NH3) ≤0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR