N-(tert-Butoxycarbonyl)-L-methionine 99+%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: BOC-L-Methionine, N-Boc-L-methionine, (tert-Butoxycarbonyl)-L-methionine, Boc-Met-OH

275630050 275630250
ACRO275630050EA 207 DKK
ACRO275630050 ACRO275630250
N-(tert-Butoxycarbonyl)-L-methionine 99+%
N-(tert-Butoxycarbonyl)-L-methionine
Formel: C₁₀H₁₉NO₄S
Smeltepunkt: 47…53 °C
Opbevaringstemperatur: Fryser
MDL Number: MFCD00065586
CAS-nummer: 2488-15-5

Bestil nu

Specifikationer Testresultater

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 49 to 55 °C
Water ≤0.5 % (K.F.)
TLC ≥99.0 %
Specific optical rotation -21.5° to -24.5° (20 °C, 589 nm) (c=1, CH3OH)
Titration with NaOH 99.0 to 101.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR