MES, (max. 1% H₂O) 99+%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2-(N-Morpholino)ethanesulphonic acid, 2-Morpholinoethanesulphonic acid

274140010 274140500 274142500
ACRO274140010EA 12800 DKK
ACRO274140010 ACRO274140500 ACRO274142500
MES, (max. 1% H₂O) 99+%
MES
Formel: C₆H₁₃NO₄S
Smeltepunkt: >300 °C (dec.)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00006181
CAS-nummer: 4432-31-9

Bestil nu

Specifikationer Testresultater

Appearance White Crystals
Infrared spectrum Conforms
Water ≤1.0 % (K.F.)
Solubility (0.5 M in water) Clear colorless
Titration with NaOH ≥99.0 % (On anhydrous basis)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR