4-Methyl-2-pentanon 99.5% for analysis

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Methylisobutylketon, Isobutylmethylketon

255660010 255665000 255660025 255660100 255660250
ACRO255660010EA 600 DKK
ACRO255660010 ACRO255665000 ACRO255660025 ACRO255660100 ACRO255660250
4-Methyl-2-pentanon 99.5% for analysis
4-Methyl-2-pentanone
Formel: (CH₃)₂CHCH₂COCH₃
Kogepunkt: 117,4 °C (760 mmHg)
Smeltepunkt: –84 °C
Massefylde: 0,8 g/cm³
Flammepunkt: 14 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008938
CAS-nummer: 108-10-1
EINECS: 203-550-1
UN nummer: 1245
ADR: 3,II
Merck Index: 13,05227
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d44947ef43cbc8103ae523ce5da380ef184ba09&vid=373b9075524004b95d32d3cf9698896714467e8c&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3950 to 1.3970 (20°C, 589 nm)
Water ≤0.2 %
GC ≥99.4 %
Color scale ≤15 APHA
Residue after evaporation ≤0.005 %
Acidity ≤0.0004 meq/g
Barium (Ba) ≤0.1 ppm
Cadmium (Cd) ≤0.05 ppm
Calcium (Ca) ≤0.5 ppm
Chromium (Cr) ≤0.02 ppm
Cobalt (Co) ≤0.02 ppm
Copper (Cu) ≤0.02 ppm
Iron (Fe) ≤0.1 ppm
Magnesium (Mg) ≤0.1 ppm
Zinc (Zn) ≤0.1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR