Kieselgel, Pore size: 40 Å 0.200-0.500 mm for chromatography

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

241670010 241672500
ACRO241670010EA 876 DKK
ACRO241670010 ACRO241672500
Kieselgel, Pore size: 40 Å 0.200-0.500 mm for chromatography
Silica gel
Formel: SiO₂
Molekylvægt: 60.08 g/mol
Kogepunkt: 2230
Smeltepunkt: 1710
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011232
CAS-nummer: 7631-86-9
REACH: 01-2119379499-16

Bestil nu

Specifikationer Testresultater

Appearance White Crystals
Particle size ≤5 % (Oversize)
Specific gravity 0.54 to 0.6 kg/l
pH 5.5 to 7 (10 % suspension in water)
Residual water 2 to 4 % (160 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR