Thionin(acetate), pure, high purity biological stain

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3,7-Diaminophenothiazin-5-ium acetate

229840050 229840250
ACRO229840050EA 569 DKK
ACRO229840050 ACRO229840250
Thionin(acetate), pure, high purity biological stain
Thionin(acetate)
Formel: C₁₄H₁₃N₃O₂S
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00081194
CAS-nummer: 78338-22-4

Bestil nu

Specifikationer Testresultater

Appearance Dark green Crystalline powder or crystals
Infrared spectrum Conforms
UV spectrum Complies
Dye content ≥85 %
UV Lambda max 597 to 601 nm (in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR