Neutral rød, pure, high purity biological stain

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: N2,N2,7-Trimethylphenazine-2,8-diamine hydrochloride, C.I. 50040, 3-Amino-7-dimethyl amino-2-methylphenazine hydrochloride, N8,N8,3-Trimethyl-2,8-phenazinediamine monohydrochloride, Toluylene red, Basic Red 5

229810250 229811000
ACRO229810250EA 865 DKK
ACRO229810250 ACRO229811000
Neutral rød, pure, high purity biological stain
Neutral rød
Formel: C₁₅H₁₇ClN₄
Smeltepunkt: 290 °C (dec.)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00012651
CAS-nummer: 553-24-2
EINECS: 209-035-8
Merck Index: 12,06571

Bestil nu

Specifikationer Testresultater

Appearance Dark green Crystalline powder
Infrared spectrum Conforms
Dye content ≥50 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR