L-Methionine methyl ester hydrochloride 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: H-Met-OMe.HCl, H-Met-OMe·HCl

229350250 229350500
ACRO229350250EA 1600 DKK
ACRO229350250 ACRO229350500
L-Methionine methyl ester hydrochloride 98%
L-Methionine methyl ester hydrochloride
Formel: C₆H₁₃NO₂S·HCl
Smeltepunkt: 151…153 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00012491
CAS-nummer: 2491-18-1
EINECS: 219-651-9

Bestil nu

Specifikationer Testresultater

Appearance White to beige Powder
Infrared spectrum Conforms
Titration Argentometric 97.5 to 102.5 %
Solubility (5 % in methanol) Clear colorless to light yellow
Specific optical rotation +24° to +28° (20°C, 589 nm) (c=1, water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR