3,3',5,5'-Tetramethylbenzidin 99+%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine

229280050 229280010 229280250
ACRO229280050EA 2340 DKK
ACRO229280050 ACRO229280010 ACRO229280250
3,3',5,5'-Tetramethylbenzidin 99+%
3,3',5,5'-Tetramethylbenzidin
Formel: C₁₆H₂₀N₂
Molekylvægt: 240.35 g/mol
Kogepunkt: 402
Smeltepunkt: 167…170 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007748
CAS-nummer: 54827-17-7
EINECS: 259-364-6

Bestil nu

Specifikationer Testresultater

Appearance White to cream to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 167 to 172 °C
Loss on drying ≤0.5 %
Sulfated ash ≤0.2 %
HPLC ≥99.0 %
Titration with HClO4 98.0 to 102.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR