Strontiumnitrat 99+% for analysis

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

222560010 222562500
ACRO222560010EA 991 DKK
ACRO222560010 ACRO222562500
Strontiumnitrat 99+% for analysis
Strontium nitrate
Formel: Sr(NO₃)₂
Kogepunkt: 645 °C (1013 hPa)
Smeltepunkt: 570 °C
Massefylde: 2,113 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011248
CAS-nummer: 10042-76-9
EINECS: 233-131-9
UN nummer: 1507
ADR: 5.1,III
Merck Index: 13,08926

Bestil nu

Specifikationer Testresultater

Appearance White Crystalline powder or crystals and/or chunks
Titration Complexometric ≥99.0 %
Loss on drying ≤0.1 % (105°C)
pH 5 to 7 (5 % at 20°C)
Chloride (Cl) ≤10 ppm
Sulfate (SO4) ≤50 ppm
Barium (Ba) ≤100 ppm
Copper (Cu) ≤2 ppm
Iron (Fe) ≤5 ppm
Magnesium (Mg) ≤20 ppm
Potassium (K) ≤50 ppm
Sodium (Na) ≤50 ppm
Heavy metals (as Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR