meta-Phosphorsyre stabilised

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Metaphosphoric acid

219221000 219225000
ACRO219221000EA 607 DKK
ACRO219221000 ACRO219225000
meta-Phosphorsyre stabilised
meta-Phosphorsyre
Formel: (HPO₃)n
Molekylvægt: 79.98 g/mol
Kogepunkt: 260
Smeltepunkt: 21
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011351
CAS-nummer: 37267-86-0
EINECS: 253-433-4
UN nummer: 3260
ADR: 8 III

Bestil nu

Specifikationer Testresultater

Appearance Colorless to white Lumps
Stabilizer 54 to 60 % NaPO3
Titration with NaOH 40 to 46 %
Chloride (Cl) ≤20 ppm
Arsenic (As) ≤5 ppm
Iron (Fe) ≤100 ppm
Heavy metals (as Pb) ≤50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR