Calciumsulfat, Drierite™ 6 mesh for drying, without indicator

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Calcium sulphate (anhydrous) anhydrite, Gypsum

219040020 219045000 219040100
ACRO219040020EA 1330 DKK
ACRO219040020 ACRO219045000 ACRO219040100
Calciumsulfat, Drierite™ 6 mesh for drying, without indicator
Calcium sulphate
Formel: CaO₄S
Smeltepunkt: 1450 °C
Massefylde: 2,964 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00010912
CAS-nummer: 7778-18-9
EINECS: 231-900-3
Merck Index: 13,01711

Bestil nu

Specifikationer Testresultater

Appearance White to greyish-white Irregular granules
Water ≤0.3 %
Particle size 6 mesh
Composition ≥99.5 % (CaSO4) Typical


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR