(R)-(-)-2-Butanol 99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: (R)-(-)-Ethyl methyl carbinol, (R)-(-)-sec-Butyl alcohol, (R)-(-)-2-Hydroxy butane, (R)-(-)-sec-Butanol, (R)-(-)-Methyl ethyl carbinol

217230010 217230050 217230250
ACRO217230010EA 528 DKK
ACRO217230010 ACRO217230050 ACRO217230250
(R)-(-)-2-Butanol 99%
(R)-(-)-2-Butanol
Formel: C₄H₁₀O
Kogepunkt: 99…100 °C (760 mmHg)
Smeltepunkt: –114 °C
Massefylde: 0,8 g/cm³
Flammepunkt: 26 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00064280
CAS-nummer: 14898-79-4
EINECS: 238-967-8
UN nummer: 1120
ADR: 3,III
Merck Index: 13,01540

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3960 to 1.3980 (20°C, 589 nm)
GC ≥98.5 %
Specific optical rotation -12° to -14° (20°C, 589 nm) (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR