3-Chlor-4-fluoranilin 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

211530250 211531000
ACRO211530250EA 241 DKK
ACRO211530250 ACRO211531000
3-Chlor-4-fluoranilin 98%
3-Chlor-4-fluoranilin
Formel: C₆H₅ClFN
Molekylvægt: 145.56 g/mol
Kogepunkt: 230
Smeltepunkt: 42…47 °C
Flammepunkt: 149 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007767
CAS-nummer: 367-21-5
EINECS: 206-682-8
UN nummer: 2811
ADR: 6.1 III

Bestil nu

Specifikationer Testresultater

Appearance Beige to brown Crystalline solid (May darken on storage)
Infrared spectrum Conforms
Melting point 42 to 47 °C
GC ≥97.5 %

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR