Magnesiumhydroxid carbonat light (∼40,0 - 45,0% MgO) for analysis

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Magnesium carbonate basic, Magnesium carbonate hydroxide, Magnesium carbonate hydrated basic

211070010 211070025
ACRO211070010EA 295 DKK
ACRO211070010 ACRO211070025
Magnesiumhydroxid carbonat light (∼40,0 - 45,0% MgO) for analysis
Magnesiumhydroxid carbonat
Formel: MgCO₃ (linear)
Molekylvægt: 84.31 g/mol
Kogepunkt: 3600
Massefylde: 2.16 g/cm³
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD07371358
CAS-nummer: 39409-82-0
EINECS: 235-192-7

Bestil nu

Specifikationer Testresultater

Appearance White Powder
Assay 40.0 to 45.0 % (MgO)
Chloride (Cl) ≤700 ppm
Sulfate (SO4) ≤3000 ppm
Arsenic (As) ≤2 ppm
Calcium (Ca) ≤7500 ppm
Iron (Fe) ≤400 ppm
Heavy metals (as Pb) ≤20 ppm
Soluble matter ≤1 % (in water)
Insoluble matter ≤0.05 % (in acetic acid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR