Bis(acetonitrile)palladium(II) dichloride 99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Bis(acetonitrile)-palladium(II) chloride, Palladium(II) chloride diacetonitrile complex, Dichlorobis(acetonitrile)palladium(II)

209455000 209450050
ACRO209455000EA 718 DKK
ACRO209455000 ACRO209450050
Bis(acetonitrile)palladium(II) dichloride 99%
Bis(acetonitrile)palladium(II) dichloride

Appearance: Orange to rust brown Crystalline powder

Formel: C₄H₆Cl₂N₂Pd
Smeltepunkt: 300 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00013122
CAS-nummer: 14592-56-4
UN nummer: 2811
ADR: 6.1,III

Bestil nu

Specifikationer Testresultater

Appearance Orange to rust brown Crystalline powder
Infrared spectrum Conforms
Elemental Analysis ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR