Kobber(II)acetat monohydrat 98+%, extra pure

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Copper diacetate monohydrate, Cupric acetate monohydrate, Acetic acid copper (II) salt monohydrate, Copper(II) acetate hydrate

203121000 203125000 203120025
ACRO203121000EA 234 DKK
ACRO203121000 ACRO203125000 ACRO203120025
Kobber(II)acetat monohydrat 98+%, extra pure
Kobber(II)acetat monohydrat
Formel: (H₃CCOO)₂Cu·H₂O
Kogepunkt: 240 °C (760 mmHg)
Smeltepunkt: 115 °C
Massefylde: 1,88 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00149570
CAS-nummer: 6046-93-1
EINECS: 205-553-3
UN nummer: 3260
ADR: 8,II
Merck Index: 13,02651

Bestil nu

Specifikationer Testresultater

Appearance Blue-green to blue Crystalline powder
Infrared spectrum Conforms
Titration Iodometric ≥98.0 %
Chloride (Cl) ≤30 ppm
Sulfate (SO4) ≤100 ppm
Iron (Fe) ≤20 ppm
Insoluble matter ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR