Hydrogenperoxid 35% (w/w) in water stabilised for analysis

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

202460010 202460025 202460250 202465000 202460100 202460251
202460010.EA 437 DKK
202460010. 202460025. 202460250. 202465000. ACRO202460100 ACRO202460251
Hydrogenperoxid 35% (w/w) in water stabilised for analysis
Hydrogen peroxide
Formel: H₂O₂
Kogepunkt: ∼107 °C (1013 hPa)
Smeltepunkt: –33 °C
Massefylde: 1,135 g/cm³
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00011333
CAS-nummer: 7722-84-1
EINECS: 231-765-0
UN nummer: 2014
ADR: 5.1,II

Bestil nu

Specifikationer Testresultater

Appearance Clear solution
Heavy metals ≤10 ppm
Color scale ≤10 APHA
Stabilizer about 0.03% stabilizers
Residue after evaporation ≤0.025 % (105°C)
Titration with KMnO4 34.5 to 37 %
Chloride (Cl) ≤2 ppm
Nitrates ≤0.012 %
Phosphates ≤0.012 %
Arsenic (As) ≤3 ppm
Chromium (Cr) ≤0.2 ppm
Iron (Fe) ≤0.5 ppm

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR