D(-)-Diethyltartrat (made from unnatural tartaric acid) 99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: (2S,3S)-Diethyl 2,3-dihydroxysuccinate, (S,S)-(-)-Diethyl tartrate, (2S,3S)-(-)-Diethyl tartrate, (-)-Diethyl D-tartrate, (S,S)-(-)-Tartaric acid diethyl ester, D(-)-Tartaric acid diethyl ester

200340250 200341000 200345000
ACRO200340250EA 551 DKK
ACRO200340250 ACRO200341000 ACRO200345000
D(-)-Diethyltartrat (made from unnatural tartaric acid) 99%
D(-)-Diethyltartrat
Formel: C₈H₁₄O₆
Molekylvægt: 206.2 g/mol
Kogepunkt: 280 °C (760 mmHg)
Smeltepunkt: 17 °C
Massefylde: 1.2 g/cm³
Flammepunkt: 93 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00064451
CAS-nummer: 13811-71-7
EINECS: 237-458-8

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4450 to 1.4470 (20°C, 589 nm)
GC ≥98.5 %
Specific optical rotation -7.5° to -9° (20°C, 589 nm) (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR