Kviksølv(II)oxid, red 99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

199991000 199995000
ACRO199991000EA 537 DKK
ACRO199991000 ACRO199995000
Kviksølv(II)oxid, red 99%
Kviksølv(II)oxid
Formel: HgO
Kogepunkt: 356,6 °C (1013 hPa)
Smeltepunkt: 500 °C (dec.)
Massefylde: 11 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011045
CAS-nummer: 21908-53-2
EINECS: 244-654-7
UN nummer: 1641
ADR: 6.1,II
Merck Index: 13,05908

Bestil nu

Specifikationer Testresultater

Appearance Orange to orange-red Powder
Titration Complexometric ≥98.5 %
Chloride (Cl) ≤500 ppm
Sulfate (SO4) ≤150 ppm
Iron (Fe) ≤200 ppm
Residue after reduction ≤0.1 %
Nitrogen compounds (as N) ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR