Praseodym(III,IV)oxid 99.9% (trace metals basis)

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

194620500
ACRO194620500EA 805 DKK
ACRO194620500
Praseodym(III,IV)oxid 99.9% (trace metals basis)
Praseodymium(III,IV) oxide
Formel: O₁₁Pr₆
Smeltepunkt: 2012…2072 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011178
CAS-nummer: 12037-29-5
EINECS: 234-857-9

Bestil nu

Specifikationer Testresultater

Appearance Dark green to dark brown to black Powder
Total conc. of imp.as REO ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR