Nikkeldichlorid hexahydrat 99.999% (trace metals basis)

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Nickel dichloride hexahydrate

193570050 193570250 193571000
ACRO193570050EA 677 DKK
ACRO193570050 ACRO193570250 ACRO193571000
Nikkeldichlorid hexahydrat 99.999% (trace metals basis)
Nikkeldichlorid hexahydrat
Formel: NiCl₂·6H₂O
Smeltepunkt: 140 °C
Massefylde: 1,84 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00149809
CAS-nummer: 7791-20-0
EINECS: 231-743-0
UN nummer: 3288
ADR: 6.1,III
Merck Index: 13,06529

Bestil nu

Specifikationer Testresultater

Appearance Green Crystalline powder or crystals and/or chunks
Total trace metal impurities ≤0.001 %
Nickel (Ni) 24 to 26 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR