Lutetium(III)oxid 99.99% (trace metals basis) -325 mesh

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Lutetium oxide, Dilutetium trioxide

193400010
ACRO193400010EA 637 DKK
ACRO193400010
Lutetium(III)oxid 99.99% (trace metals basis) -325 mesh
Lutetium(III) oxide
Formel: Lu₂O₃
Smeltepunkt: 2510 °C
Massefylde: 9,42 g/cm³
MDL Number: MFCD00011100
CAS-nummer: 12032-20-1
EINECS: 234-764-3

Bestil nu

Specifikationer Testresultater

Appearance White Powder
Assay ≥99.94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR