Sudan red 7B (fat red bluish, solvent red 19, fat red 7B)

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: N-Ethyl-1-((4-(phenyldiazenyl)phenyl)diazenyl)naphthalen-2-amine

191630050 191630500
ACRO191630050EA 514 DKK
ACRO191630050 ACRO191630500
Sudan red 7B (fat red bluish, solvent red 19, fat red 7B)
Sudan red 7B (fat red bluish, solvent red 19, fat red 7B)
Formel: C₂₄H₂₁N₅
Smeltepunkt: 130 °C (dec.)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003904
CAS-nummer: 6368-72-5
EINECS: 228-862-5

Bestil nu

Specifikationer Testresultater

Appearance Bordeaux Crystalline powder
Infrared spectrum Conforms
UV spectrum Complies


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR