4,4'-Dimethoxybenzil 99+%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Bis(4-methoxyphenyl)ethanedione, Anisil, p-Anisil, 1,2-Bis(4-methoxyphenyl)ethane-1,2-dione

170450050 170450250
ACRO170450050EA 386 DKK
ACRO170450050 ACRO170450250
4,4'-Dimethoxybenzil 99+%
4,4'-Dimethoxybenzil
Formel: C₁₆H₁₄O₄
Kogepunkt: 215…219 °C (3 mmHg)
Smeltepunkt: 131…134 °C
Massefylde: 1,36 g/cm³ (20 °C)
MDL Number: MFCD00008405
CAS-nummer: 1226-42-2
EINECS: 214-960-5

Bestil nu

Specifikationer Testresultater

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 131 to 134 °C
GC ≥99.0 %
Residue after ignition ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR