ω-Brom-4-methylacetophenon 97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: α-Bromo-4-methylacetophenone, Bromomethyl p-methylphenyl ketone, 2-Bromo-4'-methylacetophenone, 2-Bromo-1-(p-tolyl)ethanone, 4-Methylphenacyl bromide

170390050 170390250 170391000
ACRO170390050EA 358 DKK
ACRO170390050 ACRO170390250 ACRO170391000
ω-Brom-4-methylacetophenon 97%
ω-Brom-4-methylacetophenon
Formel: C₉H₉BrO
Kogepunkt: 105 °C (0,1 -rr)
Smeltepunkt: 52…53 °C
Flammepunkt: >110 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00000203
CAS-nummer: 619-41-0
EINECS: 210-595-0

Bestil nu

Specifikationer Testresultater

Appearance White to light yellow Crystalline powder or needles
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR