Thionylchlorid 99.5+%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Thionyl dichloride

169490010 169490250 169491000 169495000 169490025
ACRO169490010EA 488 DKK
ACRO169490010 ACRO169490250 ACRO169491000 ACRO169495000 ACRO169490025
Thionylchlorid 99.5+%
Thionylchlorid
Formel: Cl₂OS
Kogepunkt: 76 °C (1013 hPa)
Smeltepunkt: –105 °C
Massefylde: 1,64 g/cm³
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011449
CAS-nummer: 7719-09-7
EINECS: 231-748-8
UN nummer: 1836
ADR: 8,I
Merck Index: 12,09485

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≤0.5 % Sulfuryl chloride
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR