Anisol 99%, pure

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Methyl phenyl ether, Methoxybenzene

153920010 153920050 153922500 153920025
ACRO153920010EA 613 DKK
ACRO153920010 ACRO153920050 ACRO153922500 ACRO153920025
Anisol 99%, pure
Anisol
Formel: C₇H₈O
Kogepunkt: 154 °C (760 mmHg)
Smeltepunkt: –37 °C
Massefylde: 0,99 g/cm³
Flammepunkt: 45 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008354
CAS-nummer: 100-66-3
EINECS: 202-876-1
UN nummer: 2222
ADR: 3,III
Merck Index: 13,00672
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d32206d4bb9dbd12360d11d39b31237d54a8dcf8&vid=286a07a5e34a1ec55ff6339785af95d9a4326825&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5160 to 1.5190 (20°C, 589 nm)
Water ≤0.1 % (K.F.)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR