DL-Threonin 99.5%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

138940250 138941000 138945000
ACRO138940250EA 376 DKK
ACRO138940250 ACRO138941000 ACRO138945000
DL-Threonin 99.5%
DL-Threonin
Formel: C₄H₉NO₃
Smeltepunkt: 244 °C (dec.)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00063722
CAS-nummer: 80-68-2
EINECS: 201-300-6

Bestil nu

Specifikationer Testresultater

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Loss on drying ≤0.2 % (1 g, 105°C, 3 hrs)
Sulfated ash ≤0.1 %
Titration with HClO4 98.0 to 101.0 % (On dried substance)
Chloride (Cl) ≤200 ppm
Arsenic (As) ≤2 ppm
Heavy metals (as Pb) ≤10 ppm
Other amino acids (TLC) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR