Kulstof aktiveret, NORIT® A SUPRA

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Carbon activated, Activated carbon, Activated charcoal

134342500 134340010
ACRO134342500EA 603 DKK
ACRO134342500 ACRO134340010
Kulstof aktiveret, NORIT® A SUPRA
Kulstof aktiveret
Formel: C
Smeltepunkt: 3652 °C
Massefylde: 2 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00133992
CAS-nummer: 7440-44-0
ADR: 4.2,III

Bestil nu

Specifikationer Testresultater

Appearance Black Powder
Loss on drying ≤15 %
Sulfated ash ≤5 %
Additional info fluorescent substances passes test
Acidity/alkalinity passes test
Copper (Cu) ≤25 ppm
Lead (Pb) ≤10 ppm
Zinc (Zn) ≤25 ppm
Acid soluble substances ≤3 %
Alcohol soluble substances ≤0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR