α-Hydroxy-o-tolyl β-D-glucopyranosid 99+%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-(Hydroxymethyl)phenyl-β-D-glucopyranoside, Salicoside, D(-)-Salicin, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(2-(hydroxymethyl)phenoxy)tetrahydro-2H-pyran-3,4,5-triol, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-[2-(hydroxymethyl)phenoxy]oxane-3,4,5-triol, D(−)-Salicin

132590250 132591000 132590050
ACRO132590250EA 1930 DKK
ACRO132590250 ACRO132591000 ACRO132590050
α-Hydroxy-o-tolyl β-D-glucopyranosid 99+%
α-Hydroxy-o-tolyl β-D-glucopyranosid
Formel: C₁₃H₁₈O₇
Smeltepunkt: 197…200 °C
Massefylde: 1,42 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00006590
CAS-nummer: 138-52-3
EINECS: 205-331-6
Merck Index: 12,08476

Bestil nu

Specifikationer Testresultater

Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤0.5 %
HPLC ≥99.0 %
Specific optical rotation -59.5° to -63.5° (20°C, 589 nm) (c=5, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR