2-Phenyl-2-propanol 99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: α,α-Dimethylbenzyl alcohol

130830050 130830250 130831000
ACRO130830050EA 226 DKK
ACRO130830050 ACRO130830250 ACRO130831000
2-Phenyl-2-propanol 99%
2-Phenyl-2-propanol
Formel: C₆H₅C(OH)(CH₃)₂
Kogepunkt: 88…90 °C (11 mmHg)
Smeltepunkt: 32…34 °C
Massefylde: 0,973 g/cm³
Flammepunkt: 88 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00004456
CAS-nummer: 617-94-7
EINECS: 210-539-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f8edbb976395240a99a4f52addfbfe32e8c926f&vid=5b4ae0c8b87b317b3d1326768c6ff8bdaaab37f6&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless to white to yellow Liquid or solid
Infrared spectrum Conforms
Refractive index 1.5190 to 1.5220 (20°C, 589 nm)
GC ≥98.5 %
UV at 335 nm A: ≤1.0
Color scale ≤15 APHA
Residue after evaporation ≤0.001 %
Acidity/alkalinity ≤0.0005 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR