4,4'-Bis(dimethylamino)diphenylmethan 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 4,4'-Methylenebis(N,N-dimethylaniline), Bis[4-(dimethylamino)phenyl]methane

126740050 126741000 126745000
ACRO126740050EA 263 DKK
ACRO126740050 ACRO126741000 ACRO126745000
4,4'-Bis(dimethylamino)diphenylmethan 98%
4,4'-Bis(dimethylamino)diphenylmethan
Formel: C₁₇H₂₂N₂
Smeltepunkt: 86…90 °C
Flammepunkt: 178 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008317
CAS-nummer: 101-61-1
EINECS: 202-959-2
UN nummer: 3077
ADR: 9,III

Bestil nu

Specifikationer Testresultater

Appearance White to beige to grey to brown Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 86 to 92 °C
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR