Salpetersyre , 70% in water, pure

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Nitric acid solution

124650010 124650025 124650011 124650026
ACRO124650010EA 292 DKK
ACRO124650010 ACRO124650025 124650011. 124650026.
Salpetersyre , 70% in water, pure
Salpetersyre
Formel: HNO₃
Kogepunkt: 122 °C (760 mmHg)
Smeltepunkt: –42 °C
Massefylde: 1,4 g/cm³
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011349
CAS-nummer: 7697-37-2
UN nummer: 2031
ADR: 8,II
New Product

Bestil nu

Specifikationer Testresultater

Appearance Clear liquid (Free from suspended matter or sediment (May darken on storage))
Color scale ≤30 APHA
Titration with NaOH 67 to 70 %

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR