8-Hydroxyquinoline-5-sulphonic acid monohydrate 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 8-Hydroxyquinoline-5-sulphonic acid hydrate

121920050 121921000 121925000 121920025
ACRO121920050EA 263 DKK
ACRO121920050 ACRO121921000 ACRO121925000 ACRO121920025
8-Hydroxyquinoline-5-sulphonic acid monohydrate 98%
8-Hydroxyquinoline-5-sulphonic acid monohydrate
Formel: C₉H₇NO₄S·1H₂O
Smeltepunkt: >300 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00149482
CAS-nummer: 283158-18-9
EINECS: 201-570-5

Bestil nu

Specifikationer Testresultater

Appearance Yellow Powder
Infrared spectrum Conforms
Assay ≥97.5 %
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR