1,2-Dimethoxyethan 99+% stabilised, extra pure

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Ethylenglycoldimethylether

Stabilization: stabiliseret med BHT/Ionol

118450010 118450025 118455000 118450250
ACRO118450010EA 1030 DKK
ACRO118450010 ACRO118450025 ACRO118455000 ACRO118450250
1,2-Dimethoxyethan 99+% stabilised, extra pure
1,2-Dimethoxyethane
Formel: C₄H₁₀O₂
Kogepunkt: 84…86 °C (760 mmHg)
Smeltepunkt: –69 °C
Massefylde: 0,867 g/cm³
Flammepunkt: –6 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008502
CAS-nummer: 110-71-4
EINECS: 203-794-9
UN nummer: 2252
ADR: 3,II
Merck Index: 13,03251

Bestil nu

Specifikationer Testresultater

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3785 to 1.3815 (20°C, 589 nm)
Water ≤0.1 % (K.F.)
GC ≥99.0 %
Color scale ≤10 APHA
Stabilizer 40 to 100 ppm BHT
Peroxide value ≤15 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR