4-(Dimethylamino)cinnamaldehyde 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: p-Dimethylamino-Cinnamaldehyde

115790050 115790250
ACRO115790050EA 843 DKK
ACRO115790050 ACRO115790250
4-(Dimethylamino)cinnamaldehyde 98%
4-(Dimethylamino)cinnamaldehyde
Formel: C₁₁H₁₃NO
Smeltepunkt: 137…142 °C
Opbevaringstemperatur: Fryser
MDL Number: MFCD00007002
CAS-nummer: 6203-18-5
EINECS: 228-267-0

Bestil nu

Specifikationer Testresultater

Appearance Yellow to orange Powder
Infrared spectrum Conforms
Melting point 137 to 142 °C
Titration with HClO4 ≥97.5 %
Stabilizer 0.1 % Hydroquinone


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR