Cyclohexan 99+%, pure

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Hexahydrobenzene, Hexamethylene

111110010 111110025 111110250 111110050 111110100
ACRO111110010EA 598 DKK
ACRO111110010 ACRO111110025 ACRO111110250 ACRO111110050 ACRO111110100
Cyclohexan 99+%, pure
Cyclohexane
Formel: C₆H₁₂
Kogepunkt: 81 °C (1013 hPa)
Massefylde: 0,77 g/cm³
Flammepunkt: –18 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003814
CAS-nummer: 110-82-7
EINECS: 203-806-2
UN nummer: 1145
ADR: 3,II
Merck Index: 13,02752
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1e51ba998bef126804c3fd25b361be72cd9545&vid=d1e7f75d9f2a944eb15a7740c5254588cc9a4e67&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless Liquid
Water ≤0.05 % (K.F.)
GC ≥99.0 %
Residue after evaporation ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR