Methylthymol Blue natriumsalt, soluble_water, pure

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Sodium 2,2',2'',2'''-((((1,1-dioxido-3H-benzo[c][1,2]oxathiole-3,3-diyl)bis(6-hydroxy-5-isopropyl-2-methyl-3,1-phenylene))bis(methylene))bis(azanetriyl))tetraacetate, Methylthymol Blue tetrasodium salt

106370050
ACRO106370050EA 1270 DKK
ACRO106370050
Methylthymol Blue natriumsalt, soluble_water, pure
Methylthymol Blue natriumsalt
Formel: C₃₇H₄₀N₂Na₄O₁₃S
Molekylvægt: 844.76 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00148905
CAS-nummer: 1945-77-3
EINECS: 217-743-3

Bestil nu

Specifikationer Testresultater

Appearance Dark green to red-brown to black Powder
Infrared spectrum Conforms
Loss on drying ≤10 % (110°C, 1 h)
UV Lambda max 435 to 443 nm (in water)
Specific absorbance A (1%/1cm) ≥120 (at lambda max, in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR