Anti-WDR82 Rabbit Polyclonal Antibody

Leverandør: Abgent
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

AP4812A
ABGEAP4812AEA 2650 DKK
ABGEAP4812A
Anti-WDR82 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: WDR82 (WD repeat domain 82)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR