Anti-OR51Q1 Rabbit Polyclonal Antibody

Leverandør: Abgent
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

AP12319A
ABGEAP12319AEA 2650 DKK
ABGEAP12319A
Anti-OR51Q1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: OR51Q1 (olfactory receptor, family 51, subfamily Q, member 1 (gene/pseudogene))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR